Assay Details
Target Gene Details
Entrez Gene ID: | 13095 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 29 |
Gene Aliases: |
AHOH, AHOHase, AI159681, AI196010, Ah-2, Ahh-1, Cyp2c, P450-2C |
Location: |
Chr.19:39287044-39330713 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c29 | NM_007815.3 | NP_031841.3 | ||
| AI182545.1 | ||||
| AK149558.1 | BAE28959.1 | |||
| BC019908.1 | AAH19908.1 | |||
| D17674.1 | BAA04555.1 |
More Information
Set Membership: |
|

Genomic Map