Assay Details
Target Gene Details
Entrez Gene ID: | 329003 |
Gene Name: | zinc finger protein 516 |
Gene Aliases: |
C330029B10Rik, D230016L03, Zfp26l, Znf516, mKIAA0222 |
Location: |
Chr.18:82910137-83005314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp516 | NM_001177464.1 | NP_001170935.1 | ||
| NM_183033.2 | NP_898854.1 | |||
| AK084274.1 | ||||
| AK129092.1 | BAC97902.1 | |||
| BC053104.1 | AAH53104.1 |
More Information
Set Membership: |
|

Genomic Map