Assay Details
Target Gene Details
Entrez Gene ID: | 226089 |
Gene Name: | RAB6A GEF complex partner 1 |
Gene Aliases: |
C030046E11Rik, C130057E09Rik, Kiaa1432 |
Location: |
Chr.19:29522282-29605921 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ric1 | NM_001081319.1 | NP_001074788.1 | ||
| AK156653.1 | 12 | 1571 | BAE33795.1 | |
| AK173171.1 | BAD32449.1 |

Genomic Map