Assay Details
Target Gene Details
Entrez Gene ID: | 240665 |
Gene Name: | cyclin J |
Gene Aliases: |
D430039C20Rik |
Location: |
Chr.19:40831279-40848572 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnj | NM_172839.4 | NP_766427.1 | ||
| AK052506.1 | BAC35020.1 | |||
| AK157841.1 | BAE34222.1 | |||
| BC117559.1 | AAI17560.1 | |||
| BC120921.2 | ||||
| BC120922.1 |
More Information
Set Membership: |
|

Genomic Map