Assay Details
Target Gene Details
Entrez Gene ID: | 226143 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 44 |
Gene Aliases: |
AW107714, BC034834, b2b386Clo |
Location: |
Chr.19:44005022-44029247 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c44 | NM_001001446.3 | NP_001001446.3 | ||
| NM_001167905.1 | 8 | 1334 | NP_001161377.1 | |
| AK050436.1 | 8 | 1296 | BAC34255.1 | |
| AK149451.1 | BAE28887.1 | |||
| BC024068.1 | AAH24068.1 | |||
| BC025819.1 | AAH25819.1 | |||
| BC026766.1 | AAH26766.1 | |||
| BC034834.1 | AAH34834.1 |
More Information
Set Membership: |
|

Genomic Map