Assay Details
Target Gene Details
Entrez Gene ID: | 64697 |
Gene Name: | kidney expressed gene 1 |
Gene Aliases: |
0610008P16Rik, AI316519, GS4059 |
Location: |
Chr.19:12695790-12719902 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Keg1 | NM_029550.4 | NP_083826.1 | ||
| AB028071.1 | ||||
| AK002360.1 | BAB22039.1 | |||
| AK165349.1 | ||||
| AK165462.1 | 2 | 118 | ||
| AK165515.1 | BAE38231.1 | |||
| BC010803.1 | AAH10803.1 |
More Information
Set Membership: |
|

Genomic Map