Assay Details
Target Gene Details
Entrez Gene ID: | 12015 |
Gene Name: | BCL2-associated agonist of cell death |
Gene Aliases: |
AI325008, Bbc2 |
Location: |
Chr.19:6941861-6951905 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bad | NM_001285453.1 | 3 | 343 | NP_001272382.1 |
NM_007522.3 | 3 | 860 | NP_031548.1 | |
AK029400.1 | 1 | 843 | ||
AK131666.1 | 3 | 305 | BAE20753.1 | |
AK151792.1 | 3 | 308 | BAE30693.1 | |
AK169002.1 | 3 | 305 | BAE40801.1 | |
BC006762.1 | 3 | 828 | AAH06762.1 | |
L37296.1 | 3 | 868 | AAA64465.1 |
