Assay Details
Target Gene Details
Entrez Gene ID: | 23825 |
Gene Name: | barrier to autointegration factor 1 |
Gene Aliases: |
Baf, Bcrp1, L2bp1/Baf |
Location: |
Chr.19:5364633-5366876 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Banf1 | NM_001038231.2 | NP_001033320.1 | ||
| NM_001286608.1 | NP_001273537.1 | |||
| NM_011793.3 | NP_035923.1 | |||
| NR_104482.1 | ||||
| AB025349.1 | BAA83102.1 | |||
| AK003025.1 | BAB22518.1 | |||
| AK004212.1 | BAB23223.1 | |||
| AK010257.1 | BAB26800.1 | |||
| AK011079.1 | BAB27383.1 | |||
| AK011100.1 | BAB27397.1 | |||
| AK012588.1 | BAB28337.1 | |||
| AK088896.1 | BAC40639.1 | |||
| AK168039.1 | ||||
| BC003709.1 | AAH03709.1 | |||
| BY111854.1 | ||||
| CK780850.1 |

Genomic Map