Assay Details
Target Gene Details
Entrez Gene ID: | 29866 |
Gene Name: | calcium binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.19:4081469-4087340 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabp2 | NM_001160252.1 | NP_001153724.1 | ||
NM_001160253.1 | NP_001153725.1 | |||
NM_013878.2 | 2 | 174 | NP_038906.2 | |
AF169156.1 | AAF25790.1 | |||
AF169157.1 | 2 | 202 | AAF25791.1 | |
AK007488.1 | ||||
BC107029.2 | 2 | 174 | AAI07030.1 |
