Assay Details
Target Gene Details
Entrez Gene ID: | 381218 |
Gene Name: | RIKEN cDNA 4430402I18 gene |
Gene Aliases: |
AW122618, Spata6l |
Location: |
Chr.19:28845528-28970888 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4430402I18Rik | NM_198651.2 | NP_941053.2 | ||
| AK031997.1 | BAC27642.1 | |||
| BC147044.1 | ||||
| BC147045.1 | ||||
| BC171985.1 | ||||
| BC171986.1 |

Genomic Map