Assay Details
Target Gene Details
Entrez Gene ID: | 14227 |
Gene Name: | FK506 binding protein 2 |
Gene Aliases: |
13kDa, FKBP-2, mFKBP13, mFKBP2 |
Location: |
Chr.19:6977739-6980501 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp2 | NM_001166368.1 | NP_001159840.1 | ||
| DV655383.1 |

Genomic Map