Assay Details
Target Gene Details
Entrez Gene ID: | 60425 |
Gene Name: | double C2, gamma |
Gene Aliases: |
D830013O18Rik |
Location: |
Chr.19:4003385-4007005 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Doc2g | NM_021791.3 | 10 | 1059 | NP_068563.2 |
| AB046665.1 | 9 | 1057 | BAB16686.1 | |
| BC061104.1 | 10 | 1059 | AAH61104.1 |

Genomic Map