Assay Details
Target Gene Details
Entrez Gene ID: | 226105 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 70 |
Gene Aliases: |
- |
Location: |
Chr.19:40153361-40187286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c70 | NM_145499.2 | NP_663474.2 | ||
| AK050082.1 | ||||
| AY227736.1 | AAP55509.1 | |||
| BC016494.1 | AAH16494.1 | |||
| BC022151.1 | AAH22151.1 | |||
| BC023894.1 | ||||
| BC025822.1 | AAH25822.1 | |||
| BC034831.1 | AAH34831.1 |
More Information
Set Membership: |
|

Genomic Map