Assay Details
Target Gene Details
Entrez Gene ID: | 107250 |
Gene Name: | Kazal-type serine peptidase inhibitor domain 1 |
Gene Aliases: |
AI842353, Bono1, IGFBP-rP10 |
Location: |
Chr.19:45074485-45079289 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kazald1 | NM_178929.4 | NP_849260.2 | ||
| AK030678.1 | BAC27075.1 | |||
| AK039647.1 | ||||
| AK155095.1 | BAE33042.1 | |||
| BB618180.1 | ||||
| BB871162.1 | 1 | 38 | ||
| BC115642.1 |

Genomic Map