Assay Details
Target Gene Details
Entrez Gene ID: | 107250 |
Gene Name: | Kazal-type serine peptidase inhibitor domain 1 |
Gene Aliases: |
AI842353, Bono1, IGFBP-rP10 |
Location: |
Chr.19:45074485-45079289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kazald1 | NM_178929.4 | NP_849260.2 | ||
AK030678.1 | BAC27075.1 | |||
AK155095.1 | BAE33042.1 | |||
BC115642.1 |
