Assay Details
Target Gene Details
Entrez Gene ID: | 18011 |
Gene Name: | neuralized homolog 1A (Drosophila) |
Gene Aliases: |
2410129E16Rik, AI450910, AI481072, Neu1, Neur1, Neurl, Neurl1, Nlz, Rnf67 |
Location: |
Chr.19:47178820-47259441 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neurl1a | NM_021360.4 | 1 | 250 | NP_067335.4 |
| AK046382.1 | ||||
| BC058386.1 | 1 | 75 | AAH58386.1 | |
| BC099702.1 | 1 | 250 | AAH99702.1 |
More Information
Set Membership: |
|

Genomic Map