Assay Details
Target Gene Details
Entrez Gene ID: | 226090 |
Gene Name: | endoplasmic reticulum metallopeptidase 1 |
Gene Aliases: |
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815 |
Location: |
Chr.19:29609883-29648484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ermp1 | NM_001081213.1 | NP_001074682.1 | ||
| AK050284.1 | ||||
| AK081674.1 | BAC38286.1 | |||
| AK129447.1 | BAC98257.1 | |||
| AK137206.1 | BAE23269.1 | |||
| BC031519.1 | AAH31519.1 | |||
| BC151181.1 | ||||
| BC151182.1 | ||||
| BC157914.1 |
More Information
Set Membership: |
|

Genomic Map