Assay Details
Target Gene Details
Entrez Gene ID: | 15242 |
Gene Name: | hematopoietically expressed homeobox |
Gene Aliases: |
Hex, Hex1, Hhex-rs2, Prh, Prhx |
Location: |
Chr.19:37434841-37440731 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hhex | NM_008245.3 | NP_032271.1 | ||
| AK014111.1 | BAB29163.1 | |||
| AK033664.1 | BAC28416.1 | |||
| AK149553.1 | ||||
| BC057986.1 | AAH57986.1 |
More Information
Set Membership: |
|

Genomic Map