Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcx | NM_001162946.1 | NP_001156418.1 | ||
| AK148328.1 | BAE28484.1 | |||
| AK170596.1 | BAE41902.1 | |||
| AK170871.1 | ||||
| AK172320.1 | BAE42943.1 | |||
| BC055030.1 | AAH55030.1 | |||
| BG087228.2 | ||||
| BY230152.1 | ||||
| BY293312.1 |
More Information
Set Membership: |
|

Genomic Map