Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcx | NM_001162946.1 | 5 | 584 | NP_001156418.1 |
| NM_008797.3 | 4 | 561 | NP_032823.2 | |
| AK148328.1 | 5 | 531 | BAE28484.1 | |
| AK170596.1 | 5 | 544 | BAE41902.1 | |
| AK170871.1 | 5 | 534 | ||
| AK172320.1 | 5 | 512 | BAE42943.1 | |
| BC055030.1 | 5 | 519 | AAH55030.1 | |
| BG087228.2 | 5 | 520 | ||
| L09192.1 | 4 | 537 | AAA39737.1 |
More Information
Set Membership: |
|

Genomic Map