Assay Details
Target Gene Details
Entrez Gene ID: | 68209 |
Gene Name: | ribonuclease H2, subunit C |
Gene Aliases: |
1500026D16Rik, AYP1 |
Location: |
Chr.19:5601873-5602959 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnaseh2c | NM_026616.2 | NP_080892.1 | ||
| AF346604.1 | AAO49175.1 | |||
| AK011556.1 | BAB27694.1 | |||
| AK018776.1 | BAB31401.1 | |||
| BC024333.1 | AAH24333.1 |

Genomic Map