Assay Details
Target Gene Details
Entrez Gene ID: | 72351 |
Gene Name: | protein prenyltransferase alpha subunit repeat containing 1 |
Gene Aliases: |
1700084D09Rik, 2010013L17Rik, 4930428J16Rik, AI449677 |
Location: |
Chr.19:23686898-23723092 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptar1 | NM_028208.1 | NP_082484.1 |
More Information
Set Membership: |
|

Genomic Map