Assay Details
Target Gene Details
Entrez Gene ID: | 19265 |
Gene Name: | protein tyrosine phosphatase, receptor type, C polypeptide-associated protein |
Gene Aliases: |
AI605264, CD-45-AP, LPAP, LSM-1 |
Location: |
Chr.19:4154646-4156710 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprcap | NM_016933.3 | NP_058629.2 | ||
AA265746.1 |
