Assay Details
Target Gene Details
| Entrez Gene ID: | 225888 | 
| Gene Name: | suppressor of variegation 4-20 homolog 1 (Drosophila) | 
| Gene Aliases: | AA117471, C630029K18Rik, KMT5B, Suv4-20h1 | 
| Location: | Chr.19:3764155-3818303 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 7 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Suv420h1 | NM_001167884.1 | 7 | 1106 | NP_001161356.1 | 
| NM_001167885.1 | 4 | 804 | NP_001161357.1 | |
| NM_001167886.1 | 3 | 744 | NP_001161358.1 | |
| NM_001167887.1 | 3 | 744 | NP_001161359.1 | |
| NM_001167888.1 | 4 | 804 | NP_001161360.1 | |
| NM_001167889.1 | 3 | 744 | NP_001161361.1 | |
| NM_144871.4 | 4 | 804 | NP_659120.3 | |
| AK054093.1 | 2 | 340 | BAC35652.1 | |
| AK082660.1 | 2 | 424 | BAC38565.1 | |
| AK083227.1 | 4 | 805 | BAC38817.1 | |
| AK087267.1 | 7 | 1008 | BAC39834.1 | |
| AK139255.1 | 2 | 309 | BAE23934.1 | |
| AK152179.1 | 3 | 348 | BAE31010.1 | |
| AY555192.1 | 2 | 434 | AAT00539.1 | |
| BC011214.1 | 3 | 370 | AAH11214.1 | |
| BC075709.1 | 2 | 290 | AAH75709.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic