Assay Details
Target Gene Details
Entrez Gene ID: | 69060 |
Gene Name: | pancreatic lipase |
Gene Aliases: |
1810007A24Rik, PL, PTL |
Location: |
Chr.19:58670365-58681788 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnlip | NM_026925.3 | NP_081201.2 | ||
| AY387690.1 | AAQ90020.1 | |||
| BC061061.1 | AAH61061.1 |
More Information
Set Membership: |
|

Genomic Map