Assay Details
Target Gene Details
Entrez Gene ID: | 94063 |
Gene Name: | mitochondrial ribosomal protein L16 |
Gene Aliases: |
2310039D06Rik |
Location: |
Chr.19:11770415-11774956 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl16 | NM_025606.3 | NP_079882.2 | ||
| AB049643.1 | BAB40848.1 | |||
| AK004034.1 | ||||
| AK009687.1 | ||||
| AK011971.1 | BAB27947.1 | |||
| AK165616.1 | BAE38296.1 | |||
| BC044721.1 | AAH44721.1 |
More Information
Set Membership: |
|

Genomic Map