Assay Details
Target Gene Details
Entrez Gene ID: | 18946 |
Gene Name: | pancreatic lipase related protein 1 |
Gene Aliases: |
Plrp1 |
Location: |
Chr.19:58728887-58744169 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnliprp1 | NM_018874.2 | NP_061362.1 | ||
| AF061274.1 | AAC15774.1 | |||
| AK028105.1 | BAC25750.1 | |||
| BC090985.1 | AAH90985.1 |
More Information
Set Membership: |
|

Genomic Map