Assay Details
Target Gene Details
Entrez Gene ID: | 69826 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 10 |
Gene Aliases: |
2010001N17Rik |
Location: |
Chr.19:10962289-10974725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a10 | NM_023529.2 | NP_076018.2 | ||
| AB026046.1 | BAB18758.1 | |||
| AF237918.1 | AAK37614.1 | |||
| AK008019.1 | BAB25413.1 | |||
| BC144935.1 | ||||
| BC150675.1 |
More Information
Set Membership: |
|

Genomic Map