Assay Details
Target Gene Details
Entrez Gene ID: | 226153 |
Gene Name: | progressive external ophthalmoplegia 1 (human) |
Gene Aliases: |
D19Ertd626e, PEO, Twinl |
Location: |
Chr.19:45006079-45012762 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peo1 | NM_153796.3 | NP_722491.2 | ||
| AK054159.1 | BAE20696.1 | |||
| AK146244.1 | BAE27008.1 | |||
| AY059385.1 | AAL27647.1 | |||
| BC034909.1 | AAH34909.1 | |||
| BC071195.1 | AAH71195.1 |
More Information
Set Membership: |
|

Genomic Map