Assay Details
Target Gene Details
Entrez Gene ID: | 18648 |
Gene Name: | phosphoglycerate mutase 1 |
Gene Aliases: |
2310050F24Rik, Pgam-1 |
Location: |
Chr.19:41911871-41918665 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pgam1 | NM_023418.2 | NP_075907.2 | ||
AF283667.1 | AAG13955.1 | |||
AK004921.1 | BAB23672.1 | |||
AK009905.2 | BAB26576.1 | |||
AK144681.1 | ||||
AK150716.1 | BAE29794.1 | |||
AK152443.1 | BAE31223.1 | |||
AK153202.1 | BAE31802.1 | |||
AK159305.1 | BAE34975.1 | |||
AK165410.1 | BAE38168.1 | |||
AK168949.1 | BAE40755.1 | |||
AK180741.1 | ||||
AK185826.1 | ||||
AK190024.1 | ||||
AK190271.1 | ||||
AK192318.1 | ||||
AK192835.1 | ||||
AK195142.1 | ||||
AK195515.1 | ||||
AK196598.1 | ||||
AK197386.1 | ||||
AK197934.1 | ||||
AK199467.1 | ||||
AK203493.1 | ||||
AK212756.1 | ||||
BC002241.1 | AAH02241.1 | |||
BC005661.1 | AAH05661.1 | |||
BC066844.1 | AAH66844.1 | |||
BC067412.1 | AAH67412.1 | |||
BC083090.1 | AAH83090.1 | |||
BC106139.1 | AAI06140.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
