Assay Details
Target Gene Details
Entrez Gene ID: | 226025 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 3 |
Gene Aliases: |
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2 |
Location: |
Chr.19:22138181-22989897 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
More Information
Set Membership: |
|

Genomic Map