Assay Details
Target Gene Details
Entrez Gene ID: | 14126 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 2 |
Gene Aliases: |
FcRB, Fce1b, Fcer1b, Fcrbeta, Ms4a1, fcERI |
Location: |
Chr.19:11615517-11624034 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a2 | NM_001276328.1 | NP_001263257.1 | ||
| NM_001276329.1 | NP_001263258.1 | |||
| NM_001276330.1 | NP_001263259.1 | |||
| NM_013516.2 | NP_038544.1 | |||
| AK144021.1 | BAE25661.1 | |||
| AK161121.1 | BAE36202.1 | |||
| BB847820.1 | ||||
| BC139323.1 | ||||
| BC139324.1 | ||||
| CN663232.1 | ||||
| J05019.1 | AAA37601.1 |
More Information
Set Membership: |
|

Genomic Map