Assay Details
Target Gene Details
Entrez Gene ID: | 66704 |
Gene Name: | RNA binding motif protein 4B |
Gene Aliases: |
4921506I22Rik, AI504630, AI506404, Lark2, Neosin |
Location: |
Chr.19:4755146-4765940 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm4b | NM_025717.3 | NP_079993.2 | ||
| AK006447.1 | ||||
| AK014823.1 | ||||
| AK019871.2 | ||||
| AK028347.1 | ||||
| AK032852.1 | ||||
| AK142116.1 | ||||
| AK160796.1 | ||||
| BC019488.1 | AAH19488.1 |
More Information
Set Membership: |
|

Genomic Map