Assay Details
Target Gene Details
Entrez Gene ID: | 226026 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5 |
Location: |
Chr.19:23206441-23273908 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc5 | NM_001252684.1 | NP_001239613.1 | ||
| NM_001252685.1 | NP_001239614.1 | |||
| NM_153808.2 | NP_722503.1 | |||
| AJ534941.1 | CAD59184.1 | |||
| AK122321.1 | BAC65603.3 | |||
| AK143818.1 | ||||
| AK161254.1 | ||||
| BC007174.1 | AAH07174.1 | |||
| BC038345.1 | AAH38345.1 |

Genomic Map