Assay Details
Target Gene Details
Entrez Gene ID: | 68117 |
Gene Name: | apolipoprotein O-like |
Gene Aliases: |
6720473G16Rik, 9430083G14Rik, E130106L15Rik |
Location: |
Chr.X:112311408-112372755 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apool | NM_026565.2 | 8 | 664 | NP_080841.1 |
AK012398.1 | 8 | 710 | ||
AK012804.1 | 8 | 705 | BAB28482.1 | |
AK079141.1 | 8 | 716 | ||
BC024334.1 | 8 | 664 | AAH24334.1 |
