Assay Details
Target Gene Details
Entrez Gene ID: | 12361 |
Gene Name: | calcium/calmodulin-dependent serine protein kinase (MAGUK family) |
Gene Aliases: |
DXPri1, DXRib1, LIN-2, Pals3, mLin-2 |
Location: |
Chr.X:13517080-13846784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cask | NM_001284503.1 | NP_001271432.1 | ||
| NM_001284504.1 | NP_001271433.1 | |||
| NM_001284505.1 | NP_001271434.1 | |||
| NM_009806.3 | NP_033936.2 | |||
| AK136523.1 | BAE23024.1 | |||
| BC141292.1 | ||||
| BC145620.1 | ||||
| Y17137.1 | CAA76646.1 | |||
| Y17138.1 | CAA76647.1 |
More Information
Set Membership: |
|

Genomic Map