Assay Details
Target Gene Details
Entrez Gene ID: | 245386 |
Gene Name: | transmembrane protein 255A |
Gene Aliases: |
4933417N17, 6430550H21Rik, Fam70a |
Location: |
Chr.X:38196573-38252489 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem255a | NM_001289727.1 | NP_001276656.1 | ||
| NM_001289728.1 | NP_001276657.1 | |||
| NM_172930.4 | NP_766518.3 | |||
| AK041985.1 | BAC31122.1 | |||
| AK045375.1 | BAC32335.1 | |||
| AK077178.1 | BAC36664.1 | |||
| AK078264.1 | BAC37196.1 | |||
| BC062956.1 | AAH62956.1 | |||
| BY085502.1 | ||||
| BY085503.1 |

Genomic Map