Assay Details
Target Gene Details
Entrez Gene ID: | 54638 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
AI481216, DXImx40e, Ppp1r3f, Sfc22 |
Location: |
Chr.X:7593809-7605420 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc22 | NM_138603.3 | 1 | 139 | NP_613069.3 |
AF229637.1 | 1 | 80 | AAF66951.1 | |
AK010536.1 | 1 | 57 | BAB27013.1 | |
AK039573.1 | 1 | 112 | BAC30390.1 | |
BC011195.1 | 1 | 44 | AAH11195.1 | |
BY136942.1 | 1 | 139 |
