Assay Details
Target Gene Details
Entrez Gene ID: | 54638 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
AI481216, DXImx40e, Ppp1r3f, Sfc22 |
Location: |
Chr.X:7593809-7605420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc22 | NM_138603.3 | NP_613069.3 | ||
AF229637.1 | AAF66951.1 | |||
AK010536.1 | BAB27013.1 | |||
AK039573.1 | BAC30390.1 | |||
BC011195.1 | AAH11195.1 | |||
BM936498.1 |
Target Gene Details
Entrez Gene ID: | 20371 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
JM2, scurfin, sf |
Location: |
Chr.X:7579676-7595243 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp3 | NM_001199347.1 | 14 | 3197 | NP_001186276.1 |
NM_001199348.1 | 13 | 3055 | NP_001186277.1 | |
NM_054039.2 | 13 | 3130 | NP_473380.1 | |
AF277991.1 | 13 | 3055 | AAG53605.1 | |
AF277992.1 | 13 | 3113 | AAG53606.1 | |
AK157447.1 | 14 | 3198 | BAE34089.1 | |
DQ387959.1 | 14 | 3197 |
