Assay Details
Target Gene Details
Entrez Gene ID: | 21947 |
Gene Name: | CD40 ligand |
Gene Aliases: |
CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39 |
Location: |
Chr.X:57212143-57224042 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd40lg | NM_011616.2 | NP_035746.2 | ||
| BC119225.1 | 1 | 42 | ||
| EU274582.1 | ||||
| X65453.2 | CAA46448.2 |

Genomic Map