Assay Details
Target Gene Details
Entrez Gene ID: | 66235 |
Gene Name: | eukaryotic translation initiation factor 1A, X-linked |
Gene Aliases: |
1500010B24Rik, AI426898, Eif1ay |
Location: |
Chr.X:159372195-159385699 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1ax | NM_025437.4 | NP_079713.2 | ||
| AK005185.1 | BAB23869.1 | |||
| AK007301.1 | BAB24942.1 | |||
| AK012233.1 | BAB28110.1 | |||
| AK012714.1 | BAB28428.1 | |||
| AK021283.1 | BAB32361.1 | |||
| AK077710.1 | BAC36971.1 | |||
| AK166629.1 | BAE38904.1 | |||
| AK166852.1 | BAE39071.1 | |||
| AK187388.1 | ||||
| AK207164.1 | ||||
| AK210743.1 | ||||
| AK217305.1 | ||||
| BC027284.1 | AAH27284.1 |
More Information
Set Membership: |
|

Genomic Map