Assay Details
Target Gene Details
Entrez Gene ID: | 26905 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked |
Gene Aliases: |
AA409828, AA547477, AI314668, Eif-2gx |
Location: |
Chr.X:94188709-94212651 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2s3x | NM_012010.3 | NP_036140.1 | ||
| AJ006587.1 | CAA07099.1 | |||
| AK010819.1 | BAB27202.1 | |||
| AK128949.1 | ||||
| AK146900.1 | BAE27514.1 | |||
| AK150095.1 | BAE29303.1 | |||
| AK151579.1 | BAE30519.1 | |||
| AK153568.1 | BAE32102.1 | |||
| AK165866.1 | BAE38425.1 | |||
| AK171317.1 | ||||
| AK208120.1 | ||||
| BC063755.1 | AAH63755.1 | |||
| BC139234.1 | ||||
| BC139235.1 |
More Information
Set Membership: |
|

Genomic Map