Assay Details
Target Gene Details
Entrez Gene ID: | 13205 |
Gene Name: | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked |
Gene Aliases: |
D1Pas1-rs2, Ddx3, Fin14 |
Location: |
Chr.X:13280496-13293983 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx3x | NM_010028.3 | NP_034158.1 | ||
| AK140990.1 | BAE24540.1 | |||
| AK163250.1 | BAE37257.1 | |||
| AK188456.1 | ||||
| BC150862.1 | ||||
| BC172016.1 | ||||
| L25126.1 | AAA53630.1 | |||
| Z38117.1 | CAA86261.1 |

Genomic Map