Assay Details
Target Gene Details
Entrez Gene ID: | 67028 |
Gene Name: | RIKEN cDNA 2610002M06 gene |
Gene Aliases: |
8430408J07Rik, AI876413, Chmp1b2 |
Location: |
Chr.X:107782751-107816334 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610002M06Rik | NM_025921.3 | 1 | 139 | NP_080197.2 |
| AK011297.1 | 1 | 87 | BAB27525.2 | |
| AK019383.1 | BAB31692.2 | |||
| BC016070.1 | 1 | 78 | AAH16070.2 |
Target Gene Details
Entrez Gene ID: | 213449 |
Gene Name: | family with sequence similarity 46, member D |
Gene Aliases: |
Gm377 |
Location: |
Chr.X:107792590-107872911 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam46d | NM_001271008.1 | NP_001257937.1 | ||
| AK133246.1 |
More Information
Set Membership: |
|

Genomic Map