Assay Details
Target Gene Details
Entrez Gene ID: | 171207 |
Gene Name: | Rho GTPase activating protein 4 |
Gene Aliases: |
A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131 |
Location: |
Chr.X:73894352-73911346 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap4 | NM_001162423.1 | NP_001155895.1 | ||
| NM_001162424.1 | NP_001155896.1 | |||
| NM_138630.2 | NP_619536.2 | |||
| AF443826.1 | AAL57498.1 | |||
| AK040687.1 | BAC30667.1 | |||
| AK157294.1 | BAE34037.1 | |||
| AK169865.1 | BAE41421.1 | |||
| AY183124.1 | AAO66338.1 | |||
| AY183125.1 | ||||
| AY183126.1 | ||||
| AY183127.1 | ||||
| AY183128.1 | ||||
| AY183129.1 | ||||
| AY183130.1 | ||||
| AY183131.1 | ||||
| AY183132.1 | ||||
| AY183133.1 | ||||
| BC109001.1 | AAI09002.1 | |||
| BC109002.1 | AAI09003.1 |
More Information
Set Membership: |
|

Genomic Map