Assay Details
Target Gene Details
Entrez Gene ID: | 353283 |
Gene Name: | ES cell-expressed Ras |
Gene Aliases: |
HRAS2, HRasp, Ha-Ras2, ecat5 |
Location: |
Chr.X:7924276-7928607 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eras | NM_181548.2 | NP_853526.1 | ||
| AB093573.1 | BAC76996.1 |
More Information
Set Membership: |
|

Genomic Map