Assay Details
Target Gene Details
Entrez Gene ID: | 11977 |
Gene Name: | ATPase, Cu++ transporting, alpha polypeptide |
Gene Aliases: |
MNK |
Location: |
Chr.X:106027224-106128160 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp7a | NM_001109757.2 | 6 | 1778 | NP_001103227.1 |
| NM_009726.5 | 6 | 1775 | NP_033856.3 | |
| AB007134.1 | 5 | 1614 | BAA22369.1 | |
| BC141395.1 | 6 | 1656 | ||
| U03434.1 | 6 | 1683 | AAA57445.1 | |
| U03736.1 | 5 | 1523 | AAB08487.1 | |
| U71091.1 | 6 | 1679 | AAB37301.1 |
More Information
Set Membership: |
|

Genomic Map