Assay Details
Target Gene Details
Entrez Gene ID: | 11656 |
Gene Name: | aminolevulinic acid synthase 2, erythroid |
Gene Aliases: |
ALAS, ALAS-E, ALASE, Alas-2 |
Location: |
Chr.X:150547417-150570622 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alas2 | NM_001102446.1 | NP_001095916.1 | ||
| NM_009653.3 | NP_033783.1 | |||
| AK002642.1 | BAB22254.1 | |||
| AK077610.1 | BAC36898.1 | |||
| AK165257.1 | BAE38108.1 | |||
| AK165262.1 | ||||
| AK172094.1 | BAE42823.1 | |||
| BC150870.1 | ||||
| M15268.1 | AAA37207.1 | |||
| M63244.1 | AAA91866.1 |
Target Gene Details
Entrez Gene ID: | 77622 |
Gene Name: | apurinic/apyrimidinic endonuclease 2 |
Gene Aliases: |
C430040P13Rik, ape2 |
Location: |
Chr.X:150503629-150588191 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apex2 | AK040145.1 | BAC30522.1 | ||
| AK081677.1 | BAC38287.1 |
More Information
Set Membership: |
|

Genomic Map