Assay Details
Target Gene Details
Entrez Gene ID: | 14069 |
Gene Name: | coagulation factor VIII |
Gene Aliases: |
Cf-8, Cf8, FVIII |
Location: |
Chr.X:75172715-75383525 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F8 | NM_001161373.1 | 9 | 1728 | NP_001154845.1 |
| NM_001161374.1 | 9 | 1728 | NP_001154846.1 | |
| NM_007977.2 | 9 | 1728 | NP_032003.2 | |
| BC138347.1 | 9 | 1609 | ||
| BC145245.1 | 9 | 1609 | ||
| L05573.1 | 9 | 1726 | AAA37385.1 |
More Information
Set Membership: |
|

Genomic Map