Assay Details
Target Gene Details
Entrez Gene ID: | 20371 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
JM2, scurfin, sf |
Location: |
Chr.X:7579676-7595243 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp3 | NM_001199347.1 | NP_001186276.1 | ||
NM_054039.2 | NP_473380.1 | |||
AF277992.1 | AAG53606.1 | |||
AK157447.1 | BAE34089.1 | |||
DQ387959.1 |
Target Gene Details
Entrez Gene ID: | 78185 |
Gene Name: | protein phosphatase 1, regulatory subunit 3F, opposite strand |
Gene Aliases: |
4930524L23Rik |
Location: |
Chr.X:7573600-7581016 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3fos | NR_029473.1 | 3 | 2285 | |
AK048169.1 | 3 | 2284 |
